missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ HE-PLUS Gel Trays
Click to view available options
Dimensions (L x W):
12.5 x 6 cm
6 x 6 cm
13 x 12.5 cm
Unit Size:
Pack of 2
Pack of 4
Description
- Comes in mini (6 x 6 cm), medium (12.5 x 6 cm) or large (12.5 x 13 cm) sizes
- Either 2 or 4 pcs
Specifications
Specifications
| For Use With (Equipment) | HE-plus Horizontal Electrophoresis System |
| Type | Gel Tray |
| Dimensions (L x W) | 12.5 x 6 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction