missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ HE-PLUS Gel Trays

Product Code. p-3993808
Change view
Click to view available options
Dimensions (L x W):
12.5 x 6 cm
6 x 6 cm
13 x 12.5 cm
Unit Size:
Pack of 2
Pack of 4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
15578921 13 x 12.5 cm Pack of 2
15588921 12.5 x 6 cm Pack of 2
15598921 6 x 6 cm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15578921 Supplier Hoefer™ Supplier No. HEPLUS11

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with HE-PLUS Horizontal Electrophoresis System

  • Comes in mini (6 x 6 cm), medium (12.5 x 6 cm) or large (12.5 x 13 cm) sizes
  • Either 2 or 4 pcs

Specifications

For Use With (Equipment) HE-plus Horizontal Electrophoresis System
Type Gel Tray
Dimensions (L x W) 13 x 12.5 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.