missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ HE-PLUS Casting Kits

Product Code. 15348693
Click to view available options
Type:
Mini Casting Kit
Standard Casting Kit
Unit Size:
1 set
Each
This item is not returnable. View return policy

Product Code. 15348693

Brand: Hoefer™ HEPLUS16

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Mini Casting Kit includes four gel trays (6 × 6cm), two combs (5/8 wells) and casting stand. Standard Casting Kit includes one large gel tray (12.5 × 13cm), two medium gel trays (12.5 × 6cm), four combs (14/28 wells) and casting stand.

For use with HE-PLUS Horizontal Electrophoresis System

  • Accessories and Replacement Parts for HE Systems

Specifications

For Use With (Equipment) HE-plus Horizontal Electrophoresis System
Type Mini Casting Kit
Gel Dimensions 6 x 6 cm
Includes Four gel trays (6 x 6cm), two combs (5/8-well) and casting stand
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.