Learn More
Hoefer™ Gel Seal
Product Code. 11955338
Change view
Click to view available options
Volume (English):
0.25 oz.
Unit Size:
Each
Product Code. Volume (English) unitSize
11955338 0.25 oz. Each
1 options
Product Code. 11955338 Supplier Hoefer™ Supplier No. SE6070
A sealant used very sparingly in vertical gel casting to reduce leaking
  • For use with E245 Dual Gel Caster
Specifications
Type Gel Seal
Volume (English) 0.25 oz.