Learn More
Hoefer™ Divider Plates for SE 600 Chroma, SE 600 Standard, or SE 400 Vertical Electrophoresis Units
Accessory parts for SE 600 Chroma SE 600 Standard or SE 400 Vertical Electrophoresis Units
Brand: Hoefer™ SE6102D
Code : 04
Additional Details : Weight : 0.24300kg
Description
- Notched
- 18 x 16 cm
- Glass
Specifications
SE 600 Chroma, SE 600 Standard, SE 400 Vertical Electrophoresis Unit | |
Notched Glass Plate Divider | |
Glass |