missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ PTFE-Coated Combs for Standard-Format Vertical Gel Electrophoresis Units
For use with standard-format vertical gel electrophoresis units
Brand: Cytiva SE511-15-1.0
Code : GE
Additional Details : Weight : 0.90720kg
Description
- Available in sizes that form 10, 15, 20, or 28 wells
- Most combs are available in all three thicknesses: 0.75, 1, and 1.5mm
- Form wells that are 25mm deep (except 28-well comb which forms 15mm deep wells)
- Sample volume held by each well depends on the gel thickness, well depth, and the number of wells per comb
Specifications
SE 600, SE 400 Standard Format Vertical Gel Electrophoresis Unit | |
PTFE | |
0.03 in. | |
5.7 μL | |
0.98 in. |
Comb | |
15 | |
0.1 cm | |
0.57 cm | |
2.5 cm |