missing translation for 'onlineSavingsMsg'
Learn More

Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units

Product Code. 10289444 Shop All Cytiva Products
Change view
Click to view available options
No. of Wells:
1 Prep, 1 Reference
10
15
18
5
9
Thickness (Metric):
0.07 cm
0.075 cm
0.1 cm
0.15 cm
Volume (Metric) Well:
120μL/ mm
13μL/ mm
19.5μL/ mm
2.2μL/ mm
2.9μL/ mm
4.4μL/ mm
4.8μL/ mm
5.8μL/ mm
51μL/ mm
68μL/ mm
7.2μL/ mm
9.8μL/ mm
Unit Size:
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Wells Thickness (Metric) Volume (Metric) Well unitSize
10289444 18 0.1 cm 2.9μL/ mm Each
12019847 9 0.1 cm 5.8μL/ mm Each
10041742 18 0.1 cm 2.9μL/ mm Each
12004657 1 Prep, 1 Reference 0.15 cm 120μL/ mm Each
12904830 15 0.07 cm 2.2μL/ mm Each
11549663 15 0.07 cm 2.2μL/ mm Each
12059807 1 Prep, 1 Reference 0.07 cm 51μL/ mm Each
12004797 5 0.07 cm 9.8μL/ mm Each
12084847 10 0.075 cm 9.8μL/ mm Each
11569663 15 0.15 cm 4.4μL/ mm Each
12079857 15 0.1 cm 2.9μL/ mm Each
12074697 10 0.15 cm 7.2μL/ mm Each
10209834 10 0.15 cm 7.2μL/ mm Each
12054797 5 0.15 cm 19.5μL/ mm Each
12094797 10 0.1 cm 4.8μL/ mm Each
12044797 5 0.1 cm 13μL/ mm Each
12044737 1 Prep, 1 Reference 0.1 cm 68μL/ mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 10289444 Supplier Hoefer™ Supplier No. SE211A181,0

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems

  • Lightweight
  • Virtually unbreakable
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) SE 250, SE 260, MiniVE™ Vertical Electrophoresis System
Type Comb
Material PTFE
No. of Wells 18
Thickness (English) 0.03 in.
Thickness (Metric) 0.1 cm
Volume (Metric) Well 2.9μL/ mm
Width (Metric) 0.29 cm
Depth (English) 0.51 in.
Depth (Metric) 1.3 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.