missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
£49.30 - £92.70
Specifications
For Use With (Equipment) | SE 250, SE 260, MiniVE™ Vertical Electrophoresis System |
---|---|
Type | Comb |
Material | PTFE |
Depth (English) | 0.51 in. |
Depth (Metric) | 1.3 cm |
Product Code | Brand | Thickness (English) | Thickness (Metric) | Volume (Metric) Well | No. of Wells | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Thickness (English) | Thickness (Metric) | Volume (Metric) Well | No. of Wells | Price | Quantity & Availability | ||
11549663
|
Hoefer™
SE211A-15-.75 |
0.02 in. | 0.07 cm | 2.2μL/ mm | 15 |
£92.70
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
||
12904830
|
Hoefer™
SE211A-15-.75 |
0.02 in. | 0.07 cm | 2.2μL/ mm | 15 |
£78.22
Each |
Estimated Shipment: 09-05-2024
Log in to see stock. |
||
10041742
|
Hoefer™
SE211A-18-1.0 |
0.03 in. | 0.1 cm | 2.9μL/ mm | 18 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12004657
|
Hoefer™
SE211A-R-1.5 |
0.05 in. | 0.15 cm | 120μL/ mm | 1 Prep, 1 Reference |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12019847
|
Hoefer™
SE211A-9-1.0 |
0.03 in. | 0.1 cm | 5.8μL/ mm | 9 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10289444
|
Hoefer™
SE211A-18-1,0 |
0.03 in. | 0.1 cm | 2.9μL/ mm | 18 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12059807
|
Hoefer™
SE211A-R-.75 |
0.02 in. | 0.07 cm | 51μL/ mm | 1 Prep, 1 Reference |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12004797
|
Hoefer™
SE211A-5-.75 |
0.02 in. | 0.07 cm | 9.8μL/ mm | 5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12084847
|
Hoefer™
SE211A-10-.75 |
0.03 in. | 0.075 cm | 9.8μL/ mm | 10 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
11569663
|
Hoefer™
SE211A-15-1.5 |
0.05 in. | 0.15 cm | 4.4μL/ mm | 15 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12079857
|
Hoefer™
SE211A-15-1.0 |
0.03 in. | 0.1 cm | 2.9μL/ mm | 15 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12074697
|
Hoefer™
SE211A-10-1.5 |
0.05 in. | 0.15 cm | 7.2μL/ mm | 10 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
10209834
|
Hoefer™
SE211A-10-1.5 |
0.05 in. | 0.15 cm | 7.2μL/ mm | 10 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12054797
|
Hoefer™
SE211A-5-1.5 |
0.05 in. | 0.15 cm | 19.5μL/ mm | 5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12094797
|
Hoefer™
SE211A-10-1.0 |
0.03 in. | 0.1 cm | 4.8μL/ mm | 10 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12044797
|
Hoefer™
SE211A-5-1.0 |
0.03 in. | 0.1 cm | 13μL/ mm | 5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
12044737
|
Hoefer™
SE211A-R-1.0 |
0.03 in. | 0.1 cm | 68μL/ mm | 1 Prep, 1 Reference |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
- Lightweight
- Virtually unbreakable
Specifications
SE 250, SE 260, MiniVE™ Vertical Electrophoresis System | |
PTFE | |
1.3 cm |
Comb | |
0.51 in. |