missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoefer™ Combs for SE 250, SE 260, or miniVE Vertical Electrophoresis Units
Click to view available options
No. of Wells:
1 Prep, 1 Reference
10
15
18
5
9
Thickness (Metric):
0.07 cm
0.075 cm
0.1 cm
0.15 cm
Volume (Metric) Well:
120μL/ mm
13μL/ mm
19.5μL/ mm
2.2μL/ mm
2.9μL/ mm
4.4μL/ mm
4.8μL/ mm
5.8μL/ mm
51μL/ mm
68μL/ mm
7.2μL/ mm
9.8μL/ mm
Unit Size:
Each
Description
Made from plastics that are highly resistant to acids, bases, and all commonly used buffer systems
- Lightweight
- Virtually unbreakable
Specifications
Specifications
| For Use With (Equipment) | SE 250, SE 260, MiniVE™ Vertical Electrophoresis System |
| Type | Comb |
| Material | PTFE |
| No. of Wells | 15 |
| Thickness (English) | 0.02 in. |
| Thickness (Metric) | 0.07 cm |
| Volume (Metric) Well | 2.2μL/ mm |
| Width (Metric) | 0.29 cm |
| Depth (English) | 0.51 in. |
| Depth (Metric) | 1.3 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction