missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Hoechst Janelia Fluor 646
GREENER_CHOICE

Product Code. 18707196
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18707196

Brand: Tocris Bioscience™ 6804/100U

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Fluorogenic, red-emitting DNA probe

Hoechst Janelia Fluor™ 646 is a fluorogenic red-emitting DNA probe; it preferentially stains and binds minor groove of AT-rich regions. The compound can be combined with fluorogenic green-emitting DNA probe Hoechst Janelia Fluor™ 526 for multiplexing experiments. Hoechst Janelia Fluor™ 646 can also be combined with Hoechst Janelia Fluor™ 526 to perform dual-color STED using the same depletion laser (λdep = 775 nm). Sub-diffraction spatial resolution STED imaging can be achieved using Hoechst Janelia Fluor™ 646 in E. coli cells (∽100 nM - 1 μM).

Hoechst Janelia Fluor™ 646 is a desirable alternative to large oligonucleotide-conjugated antibodies for PAINT experiments, particularly for bacterial studies. Hoechst Janelia Fluor™ 646 exhibits a quantum yield (QY) of 0.54; extinction coefficicent 152, 000; excitation maximum = 655 nm; emission maximum = 670 nm.

Fluorogenic: fluoresces only once bound to DNA, enabling hassle-free no-wash experiments. Hoechst Janelia Fluor™ 646 is suitable for multicolor microscopy experiments and for use in live cell imaging.

To measure the absorbance spectrum of Hoechst Janelia Fluo™ 646 we recommend the following solvent: TFE plus 0.1% TFA.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-(3-(Azetidin-1-ium-1-ylidene)-7-(azetidin-1-yl)-5,5-dimethyl-3,5-dihydrodibenzo[b,e]silin-10-yl)-4-((2-(2-(2-(4-(4-(5-(4-methylpiperazin-1-yl)-1 H,3 'H-[2,5 '-bibenzo[d]imidazol]-2 '-yl)phenoxy)butanamido)ethoxy)ethoxy)ethyl)carbamoyl)benzoat
CAS 2462011-60-3
Quantity 100 μg
Target Fluorescent DNA Stains
Molecular Formula C64H70N10O7Si
Purity 0.90
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.