missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hoechst 33258, TRC
Specifications
Specifications
| Chemical Name or Material | 4-[5-(4-Methyl-1-piperazinyl)[2,5'-bi-1H-benzimidazol]-2'-yl]-phenol Trihydrochloride |
| CAS | 23491-45-4 |
| Molecular Formula | C25 H24 N6 O . 3 Cl H |
| Quantity | 100 mg |
| Synonym | Hoechst 33258, p-[5-[5-(4-Methyl-1-piperazinyl)-2-benzimidazolyl]-2-benzimidazolyl]-phenol Trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazolyl]-6-(1-methyl-4-piperazinyl)benzimidazole trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazolyl]-6-(1-methyl-4-piperazyl)-benzimidazole trihydrochloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazolyl]-6-(1-methyl-4-piperazyl)benzimidazole trichloride, 2-[2-(4-Hydroxyphenyl)-6-benzimidazolyl]-6-(1-methyl-4-piperazyl)benzimidazole-3HCl, 2’-(4-Hydroxyphenyl)-5-(4-methyl-1-piperazinyl)-2,5'-bi-1H-benzimidazole 4-[5-[5-(4-Methyl-1-piperazinyl)-2-benzimidazolyl]-2-benzimidazolyl]phenol trihydrochloride, Bisbenzimide, Bisbenzimide (quenchant), Bisbenzimide trihydrochloride, H 33258 |
| InChI Formula | InChI=1S/C25H24N6O.3ClH/c1-30-10-12-31(13-11-30)18-5-9-21-23(15-18)29-25(27-21)17-4-8-20-22(14-17)28-24(26-20)16-2-6-19(32)7-3-16;;;/h2-9,14-15,32H,10-13H2,1H3,(H,26,28)(H,27,29);3*1H |
| SMILES | Cl.Cl.Cl.CN1CCN(CC1)c2ccc3[nH]c(nc3c2)c4ccc5[nH]c(nc5c4)c6ccc(O)cc6 |
| IUPAC Name | 4-[5-[5-(4-methylpiperazin-1-yl)-1H-benzimidazol-2-yl]-1H-benzimidazol-2-yl]phenol;trihydrochloride |
| Molecular Weight (g/mol) | 533.88 |
| Formula Weight | 532.1312 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?