missing translation for 'onlineSavingsMsg'
Learn More

HO-3867, MedChemExpress

Product Code. 16499586
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499586 50 mg 50mg
16442586 25 mg 25mg
16423028 10 mg 10mg
16405767 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16499586 Supplier Medchem Express Supplier No. HY100453

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress HO-3867 is a selective and potent STAT3 inhibitor and shows good antitumor activity.

HO-3867 is a selective and potent STAT3 inhibitor and shows good antitumor activity.

Specifications

Chemical Name or Material HO-3867
CAS 1172133-28-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C28H30F2N2O2
Quantity 50 mg
Solubility Information DMSO : ≥ 32 mg/mL (68.88 mM)
SMILES O=C1/C(CN(CC2=CC(C)(C)N(O)C2(C)C)C/C1=C\C3=CC=C(F)C=C3)=C/C4=CC=C(F)C=C4
Molecular Weight (g/mol) 464.55
Formula Weight 464.55
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.