missing translation for 'onlineSavingsMsg'
Learn More

HO-1/HMOX1/HSP32 Antibody (HO-1-1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 14 publications

Brand:  Novus Biologicals NBP1-97507-0.2mg

 View more versions of this product

Product Code. 18747243

  • £635.00 / 0.20mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat, Bovine, Canine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HO-1/HMOX1/HSP32
Monoclonal
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
Mouse
33 kDa
0.2 mg
Primary
Human, Mouse, Rat, Bovine, Canine
Purified
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
HO-1-1
Western Blot 1:1000, Flow Cytometry 10 ug/ml, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
P09601
HMOX1
Synthetic peptide corresponding to the sequence near the N-terminus of human HO-1.
Protein G purified
RUO
3162
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only