Learn More
HO-1/HMOX1/HSP32 Antibody (CL11669) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15122B
Product Code. 30299289
  • £558.00 / 0.10mL
Description
HO-1/HMOX1/HSP32 Monoclonal antibody specifically detects HO-1/HMOX1/HSP32 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HO-1/HMOX1/HSP32
Monoclonal
Biotin
bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32
This antibody was developed using a recombinant protein derived from P09601, with the exact immunogen sequence remaining proprietary.
0.1 mL
Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism
3162
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin)
CL11669
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified