missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-1/HMOX1/HSP32 Antibody (CL11661) [mFluor Violet 500 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21199MFV500
This item is not returnable.
View return policy
Description
HO-1/HMOX1/HSP32 Monoclonal antibody specifically detects HO-1/HMOX1/HSP32 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| HO-1/HMOX1/HSP32 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
| This antibody was developed using a recombinant protein derived from P09601, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
| 3162 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL11661 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction