missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPUL2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-37687R
This item is not returnable.
View return policy
Description
HNRNPUL2 Polyclonal antibody specifically detects HNRNPUL2 in Human, Mouse samples. It is validated for Western Blot
Specifications
| HNRNPUL2 | |
| Polyclonal | |
| Western Blot | |
| DKFZp762N1910, heterogeneous nuclear ribonucleoprotein U-like 2, heterogeneous nuclear ribonucleoprotein U-like protein 2, HNRPUL2, SAF-A2, Scaffold-attachment factor A2 | |
| This antibody maps to a region between residue 25 to 75 of human Heterogeneous Nuclear Ribonucleoprotein U-Like 2 using the numbering given in entry NP_001073027.1 (GeneID 221092). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 221092 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction