missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11760-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene can act as a tetramer and is involved in the assembly of 40S hnRNP particles. Multiple transcript variants encoding at least two different isoforms have been described for this gene.Specifications
| HNRNPC | |
| Polyclonal | |
| Unconjugated | |
| HNRNPC | |
| C1, C2, HNRNP, hnRNP C1/C2, HNRNPC, HNRPC, SNRPC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15381, 290046, 3183 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Flow Cytometry, Immunoprecipitation | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| G3V9R8, P07910, Q9Z204 | |
| HNRNPC | |
| HNRNPC Fusion Protein Ag2356 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction