Learn More
HNRNPC Rabbit, anti-Human, Clone: 4F16, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82798-3-RR-20UL
Description
The HNRNPC antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human HNRNPC. This antibody recognizes human, mouse, rat antigen. The HNRNPC antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| HNRNPC | |
| 4F16 | |
| C1, C2, HNRNP, hnRNP C1/C2, HNRNPC, HNRPC, SNRPC | |
| 20 μL | |
| 3183 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, Immunofluorescence, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |