missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPA2B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14813-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins. The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA. These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. This gene has been described to generate two alternatively spliced transcript variants which encode different isoforms.Specifications
| HNRNPA2B1 | |
| Polyclonal | |
| Unconjugated | |
| HNRNPA2B1 | |
| hnRNP A2/B1, HNRNPA2, HNRNPA2B1, HNRNPB1, HNRPA2, HNRPA2B1, HNRPB1, RNPA2, SNRPB1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3181, 362361, 53379 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A7VJC2, O88569, P22626 | |
| HNRNPA2B1 | |
| HNRNPA2B1 Fusion Protein Ag6563 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction