Learn More
Invitrogen™ hnRNP UL1 Monoclonal Antibody (EAP 4A11), eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£1376.00 - £3115.00
Specifications
| Antigen | hnRNP UL1 |
|---|---|
| Clone | EAP 4A11 |
| Concentration | 0.5 mg/mL |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
Description: This EAP 4A11 monoclonal antibody recognizes human hnRNP UL1. EAP 4A11 detects a ∽95 kDa band corresponding to hnRNP UL1 by Western Blot and stains hnRNP UL1 in the nucleus in Immunocytochemistry. Applications Reported: This EAP 4A11 antibody has been reported for use in western blot and immunocytochemistry/immunofluorescence. Applications Tested: This EAP 4A11 antibody has been tested by western blot of human cell lysates and immunocytochemistry/immunofluorescence of human cell lines. This may be used at less than or equal to 4.0 μg/ml for western blot and less than or equal to 2.5 μg/mL for immunocytochemistry/immunofluorescence.
HNRPUL1 is a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells.This gene encodes a nuclear RNA-binding protein of the heterogeneous nuclear ribonucleoprotein (hnRNP) family. This protein binds specifically to adenovirus E1B-55kDa oncoprotein. It may play an important role in nucleocytoplasmic RNA transport, and its function is modulated by E1B-55kDa in adenovirus-infected cells. Four transcript variants encoding different isoforms have been found for this gene. Another variant has also been found, but its full-length nature has not been determined.Specifications
| hnRNP UL1 | |
| 0.5 mg/mL | |
| Unconjugated | |
| Rat | |
| Human | |
| Q9BUJ2 | |
| IgG2b κ | |
| Affinity chromatography | |
| Antibody |
| EAP 4A11 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 11100 | |
| Primary | |
| 4°C, do not freeze | |
| Hnrnpul1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.