Learn More
Invitrogen™ hnRNP U Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5143907
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human MOLT-4 whole cell, human Daudi whole cell, human HEL whole cell, human U251 whole cell, rat brain tissue, rat C6 whole cell, mouse brain tissue, mouse NIH/3T3 whole cell. IHC: human breast cancer tissue, human gastric carcinoma tissue, human colorectal adenocarcinoma tissue, human esophageal squamous carcinoma tissue, human esophageal squamous carcinoma tissue, human thyroid cancer tissue, human spleen tissue, mouse brain tissue, rat brain tissue. ICC/IF: SiHa cell. Flow: HEL cell, C6 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
The protein encoded by this gene is a receptor for activated C-kinase (RACK) protein. The encoded protein has been shown to bind in vitro to activated protein kinase C beta I. In addition, this protein is a cutaneous T-cell lymphoma-associated antigen. Finally, the protein contains a bromodomain and two zinc fingers, and is thought to be a transcriptional regulator. Multiple transcript variants encoding several different isoforms have been found for this gene.
Specifications
| hnRNP U | |
| Polyclonal | |
| Unconjugated | |
| Hnrnpu | |
| AA408410; AI256620; AL024194; AL024437; AW557595; C1orf199; C86794; FLJ30202; FLJ37978; GRIP120; Heterogeneous nuclear ribonucleoprotein U; heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A); heterogenous nuclear ribonucleoprotein U; hnRNP U; HNRNPU; HNRNPU antisense RNA 1; HNRNPU-AS1; Hnrpu; NCRNA00201; nuclear matrix protein sp120; Nuclear p120 ribonucleoprotein; p120; p120 nuclear protein; pp120; rCG_20317; SAFA; SAF-A; scaffold attachment factor A; Scaffold-attachment factor A; SN1; SP120; system N1 Na+ and H+-coupled glutamine transporter; transporter protein; transporter protein; system N1 Na+ and H+-coupled glutamine transporter; U21.1; U211 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117280, 3192, 51810 | |
| -20°C | |
| Lyophilized |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| Q00839, Q8VEK3 | |
| Hnrnpu | |
| E.coli-derived human hnRNP U/p120/HNRNPU recombinant protein (Position: E262-L563). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.