missing translation for 'onlineSavingsMsg'
Learn More

hnRNP-Q Rabbit anti-Human, Mouse, Rat, Clone: 1R0G5, Novus Biologicals™

Product Code. 18326774 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326774 100 μg 100µL
18375556 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18326774 Supplier Novus Biologicals Supplier No. NBP316838100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

hnRNP-Q Monoclonal antibody specifically detects hnRNP-Q in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen hnRNP-Q
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 1R0G5
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias dJ3J17.2, Glycine- and tyrosine-rich RNA-binding protein, GRYRBP, GRY-RBPNS1-associated protein 1, heterogeneous nuclear ribonucleoprotein Q, hnRNP Q, hnRNP-Q, HNRPQ, HNRPQ1, NSAP1FLJ31626, PP68, synaptotagmin binding, cytoplasmic RNA interacting protein, Synaptotagmin-binding, cytoplasmic RNA-interacting protein
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 524-623 of human hnRNP-Q (O60506).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10492
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.