missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hnRNP M1-M4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599240
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total hnRNP M.
Proteins that directly bind to nascent RNA polymerase II transcripts, the heterogenous nuclear ribonucleoproteins (hnRNPs), play an important role in both transcript-specific packaging and alternative splicing of pre-mRNAs. A group of abundant hnRNPs, the M1-M4 proteins, appear as a cluster of four proteins. The M proteins are pre-mRNA binding proteins in vivo, and they bind avidly to poly (G) and poly (U) RNA homopolymers in vitro. M proteins are members of the ribonucleoprotein consensus sequence family of RNA-binding proteins with greatest similarity to a hypothetical RNA-binding protein from Saccharomyces cerevisiae. The M proteins also possess an unusual hexapeptide-repeat region rich in methionine and arginine residues (MR repeat motif) that resembles a repeat in the 64kDa subunit of cleavage stimulation factor, which is involved in 3'-end maturation of pre-mRNAs. Proteins immunologically related to M exist in divergent eukaryotes ranging from human to yeast.
Specifications
| hnRNP M1-M4 | |
| Polyclonal | |
| Unconjugated | |
| HNRNPM | |
| 2610023M21Rik; AA409009; CEA receptor; CEAR; Heterogeneous nuclear ribonucleoprotein M; heterogenous nuclear ribonucleoprotein M4; hnRNA-binding protein M4; hnRNP M; HNRNP3; Hnrnpm; HNRNPM3; HNRNPM4; HNRPM; Hnrpm4; HTGR1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein M; M4 protein; mKIAA4193; N-acetylglucosamine receptor 1; NAGR1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 116655, 4670, 76936 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P52272, Q62826, Q9D0E1 | |
| HNRNPM | |
| A synthesized peptide derived from human HNRNPM(Accession P52272), corresponding to amino acid residues G23-A73. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction