missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ hnRNP M Recombinant Rabbit Monoclonal Antibody (23GB4895), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

£179.00 - £485.00

Specifications

Antigen hnRNP M
Clone 23GB4895
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30088616
GREENER_CHOICE
View Documents
Invitrogen™
MA553814
100 μL
£485.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30088308
View Documents
Invitrogen™
MA553815
20 μL
£179.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

hnRNP M Recombinant Monoclonal antibody specifically detects hnRNP M in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

hnRNP M
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Mouse, Rat
P52272, Q62826, Q9D0E1
116655, 4670, 76936
IgG
Affinity chromatography
Antibody
23GB4895
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
2610023M21Rik; AA409009; CEA receptor; CEAR; heterogeneous nuclear ribonucleoprotein M; heterogenous nuclear ribonucleoprotein M4; hnRNA-binding protein M4; hnRNP M; HNRNPM; HNRNPM4; Hnrpm; Hnrpm4; HTGR1; LOW QUALITY PROTEIN: heterogeneous nuclear ribonucleoprotein M; M4 protein; mKIAA4193; N-acetylglucosamine receptor 1; NAGR1
A synthesized peptide derived from human HNRNPM (250-300AA).
Primary
-20°C
HNRNPM
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.