missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP L Antibody (4D11), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-6106F
This item is not returnable.
View return policy
Description
hnRNP L Monoclonal specifically detects hnRNP L in Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| hnRNP L | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| FLJ35509, heterogeneous nuclear ribonucleoprotein L, hnRNP L, hnRNP-L, HNRPL | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Amphibian, Bovine, Chicken, Primate | |
| Purified |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 4D11 | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| HNRNPL | |
| Human hnRNP proteins from HeLa cells purified by affinity chromatography on ssDNA agarose. | |
| 0.1 mL | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 3191 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction