missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP K Antibody (N10 P2D3*G2), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50424UV
This item is not returnable.
View return policy
Description
hnRNP K Monoclonal specifically detects hnRNP K in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| hnRNP K | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HNRNPK | |
| Ovalbumin-conjugated synthetic peptide SVKQYSGKFF (C-terminal sequence) | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| 3190 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry | |
| N10 P2D3*G2 | |
| Western Blot, Immunohistochemistry | |
| dC-stretch binding protein, FLJ41122, heterogeneous nuclear ribonucleoprotein K, hnRNP K, transformation upregulated nuclear protein, Transformation up-regulated nuclear protein, TUNPHNRPKCSBP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction