missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ hnRNP K Antibody (F45 P9 C7), Novus Biologicals™
Mouse Monoclonal Antibody
£162.00 - £386.00
Specifications
| Antigen | hnRNP K |
|---|---|
| Clone | F45 P9 C7 |
| Dilution | Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-5 μg/mL |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066982
|
Novus Biologicals™
NBP3-30893-200ug |
200 μg |
£386.00
200µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067084
|
Novus Biologicals™
NBP3-30893-50ug |
50 μg |
£162.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
hnRNP K Monoclonal antibody specifically detects hnRNP K in Chicken,Human,Mouse,Rabbit,Rat samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)Specifications
| hnRNP K | |
| Western Blot : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Chicken, Human, Mouse, Rabbit, Rat | |
| dC-stretch binding protein, FLJ41122, heterogeneous nuclear ribonucleoprotein K, hnRNP K, transformation upregulated nuclear protein, Transformation up-regulated nuclear protein, TUNPHNRPKCSBP | |
| Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with a synthetic peptide derived from the C-terminal of the hnRNP K protein and mouse myeloma Ag8563 cells. Sequence common in human, mouse, rat, rabbit and chicken | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| F45 P9 C7 | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cancer, Core ESC Like Genes, DNA Repair, Stem Cell Markers | |
| Phosphate buffered saline | |
| 3190 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title