missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hnRNP H1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599423
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total hnRNP H.
This gene encodes a member of a subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA. These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some may shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNA and is very similar to the family member HNRPF. This gene may be associated with hereditary lymphedema type I. Alternatively spliced transcript variants have been described.
Specifications
| hnRNP H1 | |
| Polyclonal | |
| Unconjugated | |
| Hnrnph1 | |
| AI642080; E430005G16Rik; Heterogeneous nuclear ribonucleoprotein H; Heterogeneous nuclear ribonucleoprotein H, N-terminally processed; heterogeneous nuclear ribonucleoprotein H1; heterogeneous nuclear ribonucleoprotein H1 (H); hnRNP H; Hnrnph; HNRNPH1; HNRPH; HNRPH1; Ratsg1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140931, 3187, 59013 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O35737, P31943, Q8VHV7 | |
| Hnrnph1 | |
| A synthesized peptide derived from human HNRNPH1(Accession P31943), corresponding to amino acid residues H172-I222. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction