Learn More
Invitrogen™ hnRNP F Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120922
Description
Positive test controls include: 22Rv1, Lovo, MCF7, BT-474, Mouse spleen. The target is usually found in the following locations: Nucleus, nucleoplasm. Immunogen sequence: MMLGPEGGEG FVVKLRGLPW SCSVEDVQNF LSDCTIHDGA AGVHFIYTRE GRQSGEAFVE LGSEDDVKMA LKKDRESMGH RYIEVFKSHR TEMDWVLKHS GPNSADSAND GFVRLRGLPF GCTKEEIVQF FSGLEIVPNG ITLPVDPEGK ITGEAFVQFA SQELAEKALG KHKERIGHRY IEVFKSSQEE VRSYSDPPLK FMSVQRPGPY DRPGTARRYI GIVKQAGLER MRPGAYSTGY GGYEEYSGLS DGYGFTTDLF GRDLSYCLSG MYDHRYGDSE.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins. The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA. These proteins are associated with pre-mRNAs in the nucleus and regulate alternative splicing, polyadenylation, and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs which have guanosine-rich sequences. This protein is very similar to the family member hnRPH. Multiple alternatively spliced variants, encoding the same protein, have been identified.
Specifications
| hnRNP F | |
| Polyclonal | |
| Unconjugated | |
| Hnrnpf | |
| 4833420I20Rik; AA407306; heterogeneous nuclear ribonucleoprotein F; Heterogeneous nuclear ribonucleoprotein F, N-terminally processed; hnRNP F; HnRNP F protein; hnRNP H; Hnrnpf; HNRNPH1; Hnrpf; mcs94 1; mcs94-1; Nucleolin-like protein mcs94-1; OK/SW-cl.23; ribonucleoprotein F; ROF | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 3185, 98758 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P52597, Q9Z2X1 | |
| Hnrnpf | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-280 of human HNRNPF (NP_001091678.1). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.