missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hnRNP F/H Monoclonal Antibody (1G11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA536290
This item is not returnable.
View return policy
Description
Perform heat mediated antigen retrieval with citrate buffer pH 6 before commencing with IHC staining protocol. Western blot detects a band of approximately 53 kDa (hnRNP F), 56 kDa (hnRNP H). Predicted molecular weight: hnRNP F = 49.8 kDa, hnRNP H = 53.9kDa.
Specifications
| hnRNP F/H | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O35737, P31943, P52597, Q9Z2X1 | |
| HNRNPF, HNRNPH1, hnrnph1.L, hnrnph1.S | |
| Proprietary Immunogen. | |
| 50 μL | |
| Primary | |
| Human, Mouse, Frog | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
| 1G11 | |
| Unconjugated | |
| HNRNPH1 | |
| 4833420I20Rik; AA407306; AI642080; E430005G16Rik; heterogeneous nuclear ribonucleoprotein F; Heterogeneous nuclear ribonucleoprotein F, N-terminally processed; Heterogeneous nuclear ribonucleoprotein H; Heterogeneous nuclear ribonucleoprotein H, N-terminally processed; heterogeneous nuclear ribonucleoprotein H1; heterogeneous nuclear ribonucleoprotein H1 (H); heterogeneous nuclear ribonucleoprotein H1 (H) L homeolog; heterogeneous nuclear ribonucleoprotein H1 L homeolog; heterogeneous nuclear ribonucleoprotein H1 (H) S homeolog; heterogeneous nuclear ribonucleoprotein H1 S homeolog; heterogeneous nuclear ribonucleoprotein H1 L homeolog; heterogeneous nuclear ribonucleoprotein H1 S homeolog; heterogeneous nuclear ribonucleoprotein H2 (H'); hnRNP F; HnRNP F protein; hnRNP H; Hnrnpf; hnrnph; hnrnph1; hnrnph1.L; hnrnph1.S; hnrnph1-a; hnrnph1-b; hnrnph2; Hnrpf; HNRPH; hnrph1; hnrph2; mcs94-1; nucleolin-like protein mcs94-1; OK/SW-cl.23; XELAEV_18017116mg; XELAEV_18021523mg | |
| Mouse | |
| Protein A | |
| RUO | |
| 3185, 3187, 432071, 446758, 59013, 98758 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction