missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP C1 + C2 Antibody (CL2596) [Alexa Fluor™ 350], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36777AF350
This item is not returnable.
View return policy
Description
hnRNP C1 + C2 Monoclonal antibody specifically detects hnRNP C1 + C2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| hnRNP C1 + C2 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| C1, C2, heterogeneous nuclear ribonucleoprotein C (C1/C2), heterogeneous nuclear ribonucleoproteins C1/C2, HNRNP, hnRNP C1/C2, hnRNPC, HNRPC, MGC104306, MGC105117, MGC117353, MGC131677, nuclear ribonucleoprotein particle C1 protein, nuclear ribonucleoprotein particle C2 protein, SNRPC | |
| This antibody was developed using a recombinant protein derived from P07910, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| 3183 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL2596 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction