missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP A2B1 Antibody (DP3B3), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB120-6102IR
This item is not returnable.
View return policy
Description
hnRNP A2B1 Monoclonal specifically detects hnRNP A2B1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Primate samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated, Knockdown Validated.
Specifications
| hnRNP A2B1 | |
| Monoclonal | |
| DyLight 755 | |
| P22626 | |
| HNRNPA2B1 | |
| Bacterially expressed human hnRNP A2 fusion protein. | |
| 0.1 mL | |
| Cancer, DNA Repair, DNA replication Transcription Translation and Splicing | |
| 3181 | |
| Human, Mouse, Rat, Bovine, Canine, Chicken, Primate | |
| IgG2a |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoassay, KnockDown | |
| DP3B3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Knockout Validated, Knockdown Validated | |
| DKFZp779B0244, FLJ22720, heterogeneous nuclear ribonucleoprotein A2/B1, heterogeneous nuclear ribonucleoprotein B1, heterogeneous nuclear ribonucleoproteins A2/B1, hnRNP A2 hnRNP B1, HNRNPA2, HNRNPB1, HNRPA2, HNRPA2B1hnRNP A2/B1, HNRPB1, nuclear ribonucleoprotein particle A2 protein, RNPA2, SNRPB1 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse, Human, Chicken, Canine, Rat, Monkey, Bovine and Hamster. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction