missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP A1 Antibody (4B10), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-672C
This item is not returnable.
View return policy
Description
hnRNP A1 Monoclonal specifically detects hnRNP A1 in Human, Mouse, Bovine, Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.
Specifications
| hnRNP A1 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HNRNPA1 | |
| Full length partially purified human hnRNP A1. [UniProt# P09651] | |
| 0.1 mL | |
| Primary | |
| Does not react with hnRNP A2 or B | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown | |
| 4B10 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated | |
| Helix-destabilizing protein, heterogeneous nuclear ribonucleoprotein A1, heterogeneous nuclear ribonucleoprotein A1B protein, heterogeneous nuclear ribonucleoprotein B2 protein, heterogeneous nuclear ribonucleoprotein core protein A1, hnRNP A1, hnRNP core protein A1, hnRNPA1, hnRNP-A1, HNRPA1MGC102835, nuclear ribonucleoprotein particle A1 protein, single-strand DNA-binding protein UP1, Single-strand RNA-binding protein | |
| Mouse | |
| Protein G purified | |
| Core ESC Like Genes, Stem Cell Markers | |
| 3178 | |
| Human, Mouse, Bovine, Canine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction