missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNMT, Mouse anti-Human, Clone: 2G24, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00003176-M12.50ug
This item is not returnable.
View return policy
Description
Sequence: KKYGSRFPQDDLCQYITSDDLTQMLDNLGLKYECYDLLSTMDISDCFIDGNENGDLLWDFLTETCNFNATAPPDLRAELGKDLQEPEFSAKKEGKVLFNNTLSFIVIEASpecifications
| HNMT | |
| Monoclonal | |
| Unconjugated | |
| histamine N-methyltransferase | |
| HMT/HNMT-S1/HNMT-S2 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 2G24 | |
| In 1x PBS, pH 7.4 | |
| NM_006895 | |
| HNMT | |
| HNMT (NP_008826, 184 a.a. ∼ 292 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 3176 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction