Learn More
Invitrogen™ HNMT Monoclonal Antibody (K40053_20E2), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA556158
Description
Sequence of this protein is as follows: PGIIGRIGDT KSEIKILSIG GGAGEIDLQI LSKVQAQYPG VCINNEVVEP SAEQIAKYKE LVAKTSNLEN VKFAWHKETS SEYQSRMLEK KELQKWDFIH MIQMLYYVKD IPATLKFFHS LLGTNAKMLI IVVSGSSGWD KLWKKYGSRF PQDDLCQYIT SDDLTQMLDN LGLKYECYDL LSTMDISDCF IDGNENGDLL WDFLTETCNF NATAPPDLRA ELGKDLQEPE FSAKKEGKVL FNNTLSFIVI
In mammals, histamine is metabolized by two major pathways: N(tau)-methylation via histamine N-methyltransferase and oxidative deamination via diamine oxidase. HNMT is the first enzyme which is found in the cytosol and uses S-adenosyl-L-methionine as the methyl donor. In the mammalian brain, the neurotransmitter activity of histamine is controlled by N(tau)-methylation as diamine oxidase is not found in the central nervous system. A common genetic polymorphism affects the activity levels of this gene product in red blood cells.
Specifications
| HNMT | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| P50135 | |
| Hnmt | |
| Recombinant human HNMT (immunogen range is 41-290). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| K40053_20E2 | |
| Unconjugated | |
| Hnmt | |
| 1500031F01Rik; AI788969; histamine N-methyltransferase; Hmt; Hnmt; HNMT-S1; HNMT-S2; MRT51 | |
| Mouse | |
| Protein A | |
| RUO | |
| 3176 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.