missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF4A Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518363
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with canine and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Hepatocyte nuclear factor 4 alpha (HNF4 alpha) is an orphan nuclear receptor. HNF4a is expressed in the liver, kidney, intestine, and pancreas. Mutation of HNF4a in humans has been associated with maturity-onset diabetes of the young type 1 (MODY1). HNF4 binds to DNA as an exclusive homodimer. The HNF4a gene is alternatively spliced and may generate up to nine different isoforms, HNF4a1 through HNF4a9.Specifications
HNF4A | |
Polyclonal | |
Unconjugated | |
HNF4A | |
alpha transcription factor 4; FLJ39654; FRTS4; hepatic nuclear factor 4 alpha; hepatic nuclear factor 4, alpha; hepatocyte nuclear factor 4 alpha; hepatocyte nuclear factor 4, alpha; hepatocyte nuclear factor 4-alpha; HNF4; Hnf-4; HNF4 alpha; Hnf4a; HNF4a7; HNF4a8; HNF4a9; Hnf4alpha; HNF-4-alpha; HNF4-alpha; HNF4alpha10; HNF4alpha10/11/12; HNF4alpha11; HNF4alpha12; MODY; MODY1; NR2A1; NR2A21; Nuclear receptor 2A1; nuclear receptor subfamily 2 group A member 1; TCF; TCF14; TCF-14; Transcription factor 14; transcription factor HNF-4 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
3172 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P41235 | |
HNF4A | |
Synthetic peptide sequence (RLSKTLVDMDMADY) corresponding to the N-terminus amino acids of HNF4A | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |