missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TCF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549690
This item is not returnable.
View return policy
Description
The antibody detects endogenous level of total HNF1B protein.
Hepatocyte Nuclear Factor 1 (HNF1, Homeoprotein LFB3, Transcription factor 2, TCF2, Variant hepatic nuclear factor) is a member of the homeodomain-containing superfamily of transcription factors. It is a liver-specific factor of the homeobox-containing basic helix-turn-helix family. The protein binds to DNA as either a homodimer, or a heterodimer with the related protein hepatocyte nuclear factor 1-alpha. HNF1has been shown to function in nephron development and regulates development of the embryonic pancreas. Mutations in the gene result in renal cysts and diabetes syndrome and noninsulin-dependent diabetes mellitus, and expression of the gene is altered in some types of cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| TCF2 | |
| Polyclonal | |
| Unconjugated | |
| HNF1B | |
| FJHN; Hepatocyte nuclear factor 1-beta; hepatocyte nuclear factor-1 beta; HNF1 beta A; HNF1 homeobox B; HNF1B; Hnf-1b; Hnf1beta; HNF-1Beta; HNF-1-beta; HNF2; homeoprotein LFB3; HPC11; LFB3; LF-B3; MODY5; Tcf2; Tcf-2; Transcription factor 2; Transcription factor 2 hepatic; Transcription factor 2 hepatic; LF-B3; variant hepatic nuclear factor; transcription factor 2, hepatic; Transcription factor 2, hepatic; LF-B3; variant hepatic nuclear factor; variant hepatic nuclear factor 1; vHNF1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 21410, 25640, 6928 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P23899, P27889, P35680 | |
| HNF1B | |
| Peptide sequence around aa.252~256(R-Q-K-N-P) derived from Human HNF-1beta(TCF-2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction