missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF1A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22426-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene is expressed ubiquitously with higher levels in fetal than in adult tissues. It encodes a protein sharing 93% sequence identity with the mouse protein. Wolf-Hirschhorn syndrome (WHS) is a malformation syndrome associated with a hemizygous deletion of the distal short arm of chromosome 4. This gene is mapped to the 165 kb WHS critical region, and may play a role in the phenotype of the WHS or Pitt-Rogers-Danks syndrome. The encoded protein is found to be capable of reacting with HLA-A2-restricted and tumor-specific cytotoxic T lymphocytes, suggesting a target for use in specific immunotherapy for a large number of cancer patients. This protein has also been shown to be a member of the NELF (negative elongation factor) protein complex that participates in the regulation of RNA polymerase II transcription elongation.Specifications
| HNF1A | |
| Polyclonal | |
| Unconjugated | |
| HNF1A | |
| HNF 1 alpha, HNF 1A, HNF1, HNF1 homeobox A, HNF1A, LFB1, MODY3, TCF 1, TCF1, Transcription factor 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21405, 24817, 6927 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.56 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15257, P20823, P22361 | |
| HNF1A | |
| HNF1A Fusion Protein Ag18188 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction