missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF1, Mouse anti-Human, Clone: HNF1A/2087, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79777-100UG
This item is not returnable.
View return policy
Description
HNF1 Monoclonal specifically detects HNF1 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| HNF1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P20823 | |
| HNF1A | |
| Recombinant fragment (around aa 214-339) of human HNF1 protein (exact sequence is proprietary) | |
| 100 μg | |
| Transcription Factors and Regulators | |
| 6927 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| HNF1A/2087 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| HNF1 homeobox A, IDDM20, LFB1transcription factor 1, hepatic; LF-B1, hepatic nuclear factor (HNF1), albuminproximal factor, MODY3, TCF1hepatic, Transcription factor 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction