missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32126PCP
This item is not returnable.
View return policy
Description
HNF1 Polyclonal antibody specifically detects HNF1 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative)
Specifications
| HNF1 | |
| Polyclonal | |
| Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative) | |
| HNF1 homeobox A, IDDM20, LFB1transcription factor 1, hepatic, LF-B1, hepatic nuclear factor (HNF1), albuminproximal factor, MODY3, TCF1hepatic, Transcription factor 1 | |
| The immunogen this antibody was made to, maps to a region between residue 581 to 631 of human HNF1 Homeobox A using the numbering given in entry NP_000536.5 (GeneID 6927). | |
| 0.1 mL | |
| Transcription Factors and Regulators | |
| 6927 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction