missing translation for 'onlineSavingsMsg'
Learn More

HNF1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18424547 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424547 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424547 Supplier Novus Biologicals Supplier No. NBP232123AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

HNF1 Polyclonal antibody specifically detects HNF1 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias HNF1 homeobox A, IDDM20, LFB1transcription factor 1, hepatic; LF-B1, hepatic nuclear factor (HNF1), albuminproximal factor, MODY3, TCF1hepatic, Transcription factor 1
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 50 to 100 of human HNF1 Homeobox A using the numbering given in entry NP_000536.5 (GeneID 6927).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 6927
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.