missing translation for 'onlineSavingsMsg'
Learn More

HNF-3 beta/FoxA2 Antibody (OTI3C10), PE/Cy7, Novus Biologicals™

Product Code. 18119622 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18119622 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18119622 Supplier Novus Biologicals Supplier No. NBP270904PECY7

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

HNF-3 beta/FoxA2 Monoclonal specifically detects HNF-3 beta/FoxA2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNF-3 beta/FoxA2
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3C10
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias forkhead box A2, Forkhead box protein A2, hepatic nuclear factor-3-beta, hepatocyte nuclear factor 3, beta, hepatocyte nuclear factor 3-beta, HNF-3B, HNF-3-beta, HNF3BTCF-3B, MGC19807, TCF3B, Transcription factor 3B
Gene Symbols FOXA2
Host Species Mouse
Immunogen Recombinant protein expressed in E.coli corresponding to amino acids 300-458 of human HNF-3 beta/FoxA2.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3170.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.