missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-3 beta/FoxA2 Antibody (OTI3C10), mFluor Violet 610 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
HNF-3 beta/FoxA2 Monoclonal antibody specifically detects HNF-3 beta/FoxA2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | HNF-3 beta/FoxA2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI3C10 |
| Conjugate | mFluor Violet 610 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | forkhead box A2, Forkhead box protein A2, hepatic nuclear factor-3-beta, hepatocyte nuclear factor 3, beta, hepatocyte nuclear factor 3-beta, HNF-3B, HNF-3-beta, HNF3BTCF-3B, MGC19807, TCF3B, Transcription factor 3B |
| Host Species | Mouse |
| Immunogen | Recombinant protein expressed in E.coli corresponding to amino acids 300-458 of human FOXA2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?