missing translation for 'onlineSavingsMsg'
Learn More

HNF-3 alpha/FoxA1 Mouse anti-Human, Rat, Clone: rFOXA1/1515, Novus Biologicals™

Mouse Monoclonal Antibody

£197.00

Specifications

Antigen HNF-3 alpha/FoxA1
Clone rFOXA1/1515
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Flow Cytometry, Immunohistochemistry (Paraffin)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18323763
View Documents
Novus Biologicals
NBP3-07177-20UG
20 μg
£197.00
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

HNF-3 alpha/FoxA1 Monoclonal specifically detects HNF-3 alpha/FoxA1 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry-Paraffin.
Specifications

Specifications

HNF-3 alpha/FoxA1
0.2 mg/ml
Flow Cytometry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Breast Cancer, Cancer, Stem Cell Markers
10 mM PBS with 0.05% BSA
3169
IgG1 κ
Protein A or G purified
rFOXA1/1515
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human, Rat
forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A
Recombinant human HNF-3 alpha/FoxA1 protein fragment (around aa372-472) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.