missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-3 alpha/FoxA1 Antibody (FOXA1/1519), Alexa Fluor™ 488, Novus Biologicals™
Shop All Bio Techne ProductsDescription
HNF-3 alpha/FoxA1 Monoclonal antibody specifically detects HNF-3 alpha/FoxA1 in Human, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
Specifications
| Antigen | HNF-3 alpha/FoxA1 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | FOXA1/1519 |
| Conjugate | Alexa Fluor 488 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A |
| Host Species | Mouse |
| Immunogen | Recombinant fragment of human HNF-3 alpha/FoxA1 protein (aa372-472) (exact sequence is proprietary) (Uniprot: P55317 ) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?