missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-3 alpha/FoxA1 Antibody (3B11NB) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£183.00 - £335.00
Specifications
| Antigen | HNF-3 alpha/FoxA1 |
|---|---|
| Clone | 3B11NB |
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 2-4 ug/ml, Flow Cytometry 25 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:25 - 1:50, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18133229
|
Novus Biologicals
NBP2-45269SS |
0.025 mg |
£183.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18139238
|
Novus Biologicals
NBP2-45269 |
0.1 mg |
£335.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
HNF-3 alpha/FoxA1 Monoclonal specifically detects HNF-3 alpha/FoxA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| HNF-3 alpha/FoxA1 | |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| PBS with 0.02% Sodium Azide | |
| forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A | |
| FOXA1 | |
| IgG2b κ | |
| Protein G purified |
| 3B11NB | |
| Western Blot 2-4 ug/ml, Flow Cytometry 25 ug/ml, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 1:25 - 1:50, Immunohistochemistry-Paraffin 5 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| P55317 | |
| 3169 | |
| Partial recombinant human FOXA1 protein (between amino acids 350-500) [Uniprot: P55317]. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title