missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-3 alpha/FoxA1 Antibody (3B11NB), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45269AF405
This item is not returnable.
View return policy
Description
HNF-3 alpha/FoxA1 Monoclonal antibody specifically detects HNF-3 alpha/FoxA1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| HNF-3 alpha/FoxA1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A | |
| Partial recombinant human FOXA1 protein (between amino acids 350-500) [Uniprot: P55317]. | |
| 0.1 mL | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| 3169 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 3B11NB | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction