missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HN1L Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559779
This item is not returnable.
View return policy
Description
Immunogen sequence: GSRAMKPPGG ESSNLFGSPE EATPSSRPNR MASNIFGPTE EPQNIPKRTN PPGGKGSGIF DESTPVQTRQ HLNPPGGKTS DI Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 87%.
Hematological and neurological expressed 1-like protein (HN1L) is expressed in liver, kidney, prostate, testis and uterus.
Specifications
| HN1L | |
| Polyclonal | |
| Unconjugated | |
| JPT2 | |
| 2810430B18Rik; AL022629; C16orf34; CRAMP_1 like; D17Ertd441e; hematological and neurological expressed 1-like; hematological and neurological expressed 1-like protein; hematopoietic- and neurologic-expressed sequence 1-like; HN1 like; HN1L; HN1-like protein; Jpt2; Jupiter microtubule associated homolog 2; L11; RGD1305117 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 90861 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H910 | |
| JPT2 | |
| Recombinant protein corresponding to Human HN1L. Recombinant protein control fragment (Product #RP-98813). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction