missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HN1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33436-100ul
This item is not returnable.
View return policy
Description
HN1 Monoclonal antibody specifically detects HN1 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| HN1 | |
| Monoclonal | |
| Western Blot 1:200 - 1:500, ELISA Recommended starting concentration is 1 μg/mL | |
| Androgen-regulated protein 2, ARM2hematological and neurological expressed 1 protein, hematological and neurological expressed 1, HN1A, OPMRX60 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 50-154 of human HN1 (NP_057269.1).,, Sequence:, NIFGTPEENQASWAKSAGAKSSGGREDLESSGLQRRNSSEASSGDFLDLKGEGDIHENVDTDLPGSLGQSEEKPVPAAPVPSPVAPAPVPSRRNPPGGKSSLVLG | |
| 100 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51155 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction