missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMX3 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-85051-0.1ML
This item is not returnable.
View return policy
Description
HMX3 Polyclonal specifically detects HMX3 in Mouse samples. It is validated for Western Blot.
Specifications
| HMX3 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 340784 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| H6 family homeobox 3, homeo box (H6 family) 3, homeobox (H6 family) 3, Homeobox protein H6 family member 3, homeobox protein HMX3, Homeobox protein Nkx-5.1, NKX5.1, NKX-5.1, NKX5-1 | |
| The immunogen is a synthetic peptide directed towards the middle region of Mouse HMX3 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction